Order Entry
Germany
ContactUsLinkComponent
 
3,5-Dinitro-o-toluamide
Catalog # EHERC12786050
Supplier:  EHRENSTORFER
CAS Number:  
3,5-Dinitro-o-toluamide
Catalog # EHERC12786050
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :225.16
Merck Index:12,03321
Melting Pt:180 °C
EINECS:205-706-4
CAS Number:148-01-6
Formula:C₈H₇N₃O₅
MDL Number:MFCD00072065