Order Entry
Germany
ContactUsLinkComponent
Sudan III
Sudan III
Catalog # EHERC16986103
Supplier:  EHRENSTORFER
CAS Number:  
Sudan III
Catalog # EHERC16986103
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :352.4 g/mol
Merck Index:12,09054
Melting Pt:205.3…206.3 °C
EINECS:201-638-4
CAS Number:85-86-9
Formula:C₂₂H₁₆N₄O
MDL Number:MFCD00003905