Sudan III
Catalog # MOLE43449981-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:352.4 g/mol
Merck Index:12,09054
Melting Pt:205.3…206.3 °C
EINECS:201-638-4
CAS Number:85-86-9
Formula:C₂₂H₁₆N₄O
MDL Number:MFCD00003905