Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sudan III
Sudan III, Sigma-Aldrich®
Sudan III, Sigma-Aldrich®
Sudan III, Sigma-Aldrich®
SIALS4131-25G
:
Merck
:
Sudan III, Sigma-Aldrich®
SIALS4131-25G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
352.4 g/mol
:
12,09054
:
205.3…206.3 °C
:
M-Clarity™ quality level = MQ200
:
201-638-4
:
85-86-9
:
C₂₂H₁₆N₄O
:
MFCD00003905