Order Entry
Germany
ContactUsLinkComponent
Sudan III, Supelco®
Sudan III, Supelco®
Catalog # 68562-25MG
Supplier:  Merck
CAS Number:  
undefined
Sudan III, Supelco®
Catalog # 68562-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :352.4 g/mol
Merck Index:12,09054
Melting Pt:205.3…206.3 °C
 :M-Clarity™ quality level = MQ100
EINECS:201-638-4
CAS Number:85-86-9
Formula:C₂₂H₁₆N₄O
MDL Number:MFCD00003905