Order Entry
Germany
Orders LinkContactUsLinkComponent
Chlorphenesin

Chlorphenesin

25 g
Catalog # FLUO003642-25G
Marketsource
Supplier:   FLUOROCHEM
Chlorphenesin
Catalog # FLUO003642-25G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  104-29-0
Chlorphenesin

25 g

CAS Number:  104-29-0
Price
63,30 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:104-29-0
  • Formula:C₉H₁₁ClO₃
  • MDL Number:MFCD00021990
  • Molecular Weight:202.64 g/mol
  • Melting Pt:80 °C
  • EINECS:203-192-6
Specifications

Specifications

  • Pk:
    25 g

Specifications