Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Nuclear Fast Red

Glass bottle, 125 mL
Catalog # DIAN15904
Marketsource
Supplier:   DIANOVA
Nuclear Fast Red
Catalog # DIAN15904
Supplier:  DIANOVA
Marketsource
CAS Number:  6409-77-4
Nuclear Fast Red

Glass bottle, 125 mL

CAS Number:  6409-77-4
Price
Inquire For Price
Log in for availability
1 * 125 mL
Product Details & Documents
Synonyms:  CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL
  • CAS Number:6409-77-4
  • Formula:C₁₄H₈NNaO₇S
  • MDL Number:MFCD00078493
  • Molecular Weight:357.28 g/mol
  • Merck Index:13,06512
  • Melting Pt:300 °C
  • Storage Temperature:Ambient
  • EINECS:229-088-0
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    125 mL

Specifications