
1,5-Dihydroxy-9,10-anthraquinone, Sigma-Aldrich®
Catalog # SIAL340014-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:240.22 g/mol
EINECS:204-175-6
:GHS07
CAS Number:117-12-4
Formula:C₁₄H₈O₄
MDL Number:MFCD00001210