Order Entry
Germany
ContactUsLinkComponent
N,N,3,5-Tetramethylaniline, Sigma-Aldrich®
N,N,3,5-Tetramethylaniline, Sigma-Aldrich®
Catalog # SIAL177342-25G
Supplier:  Merck
CAS Number:  
N,N,3,5-Tetramethylaniline, Sigma-Aldrich®
Catalog # SIAL177342-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :149.24 g/mol
Boiling Pt:88 °C (1013 hPa)
Density:0.9275 g/cm³ (25 °C)
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
EINECS:225-539-0
UN:2810
CAS Number:4913-13-7
Formula:C₁₀H₁₅N
MDL Number:MFCD00008307