Order Entry
Germany
ContactUsLinkComponent
4-Aminohippuric acid
4-Aminohippuric acid
Catalog # MOLE10722816-100G
Supplier:  Molekula
CAS Number:  
4-Aminohippuric acid
Catalog # MOLE10722816-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :194.19 g/mol
Merck Index:13,00441
Boiling Pt:441 °C (1013 hPa)
Melting Pt:199…202 °C
Density:1.402 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-518-9
CAS Number:61-78-9
Formula:H₂NC₆H₄C(O)NHCH₂COOH
MDL Number:MFCD00007890