ContactUsLinkComponent
CellAdhere™ Tropoelastin, Human, Lyophilised
CellAdhere™ Tropoelastin, Human, Lyophilised
  STMC07003
 :  
undefined
CellAdhere™ Tropoelastin, Human, Lyophilised
  STMC07003
 :  
  • Pk:
    1 mg
  • Species:
    Non-human primate
    Human
    Rat
    Mouse
  • Description:
    Tropoelastin, human, lyophilised
Purified tropoelastin, human, lyophilised, for tissue engineering research and cell culture - 1 mg
Coat cultureware with CellAdhere™ Tropoelastin, human, lyophilised to promote cell attachment and spreading, while enabling the modulation of surface elasticity, which is known to influence cell growth and function. The water soluble precursor to elastin, tropoelastin is a structural protein that provides strength and elasticity to tissues in the body. Ideal for coating surfaces for tissue engineering research and cell culture, this purified, human, lyophilised tropoelastin powder is sterile and contains a single recombinant protein composed entirely of human sequences. Endotoxin levels are 10,0 EU/ml or less, as assayed by Limulus amoebocyte lysate (LAL) assay.
 : Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').