Order Entry
Germany
ContactUsLinkComponent
 
Trypsin-EDTA (0,05%)
  STMC07910
 :  
undefined
Trypsin-EDTA (0,05%)
  STMC07910
 :  

 

  • Pk:
    500 mL
  • Species:
    Non-human primate
    Human
    Rat
    Mouse
  • Description:
    Trypsin-EDTA (0,05%)

 

 

Enzymatic cell dissociation reagent.

  • Generate single-cell suspenstions of human ES and iPS cells for downstream assays with a gentle, low concentration Trypsin-EDTA solution

Generate single-cell suspensions of human embryonic stem cells (ES) or human induced pluripotent stem (iPS) cells for flow cytometry or cloning assays with Trypsin-EDTA (0,05%). EDTA weakens cell-cell adhesion, which improves trypsin's access to hydrolysis-targeted peptide bonds, and enhances detachment of adherent cells. This is a gentler version than the standard concentration (0,25%), and has applications to many other cell culture systems in addition to human ES cells and iPS cells.

 : Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').