Specifications
- Pk:100 mL
- Species:Non-human primateHumanRatMouse
- Description:ACCUMAX™ cell detachment solution
Specifications
About this item
Cell detachment solution.
- Increase the accuracy of cell counts, using manual or automated methods
- Effectively dissolve cell clumps, such as spheroids and neurospheres
- Obtain cleaner cultures with a ready-to-use solution, free from mammalian and bacterial-derived products
Generate single-cell suspensions from clumped cell cultures for accurate cell counting with ACCUMAX™. This ready-to-use solution of proteolytic and collagenolytic enzymes effectively dissolves cell clumps and effectively facilitates the detachment of cells from primary tissue, as well as standard and adhesion-coated culture plasticware.
ACCUMAX™ is free of mammalian- or bacterial- derived products to ensure reproducible and reliable experiments, and has been shown to work effectively across a wide variety of cell types. Each lot of ACCUMAX™ is tested for sterility (by USP membrane filtration method), enzymatic activity (tested with synthetic chromogenic tetrapeptides), and cell detachment from tissue culture plastic.