Collagenase Type II
Supplier: STEMCELL Technologies
Certificates
About this item
For digestion of native collagen fibrils.
- Use in a variety of cell and tissue types
Digest native collagen fibrils in connective tissues with Collagenase Type II, sourced from Clostridium histolyticum. Collagenase preparations contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler and Yaron). With high levels of protease activity, especially clostripain, Collagenase Type II has been used for the digestion of many tissues and cell types, including lung (You and Brody; Comhair et al.), kidney (Valente et al.), liver (Linghor et al.), thymus (Vandenabeele et al.), endothelial cells (Patel et al.), and mesenchymal stem cells (Steigman and Fauza).
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
- Species:Non-human primateHumanRatMouse