Specifications
- Pk:2 mL
- Species:Human
- Description:Vitronectin XF™
- Seq:01
Specifications
About this item
Defined, xeno-free matrix that supports the growth and differentiation of human pluripotent stem cells under serum-free, feeder-free conditions.
- Decrease sources of variability in experiments with a recombinant human protein matrix
- Handle at room temperature without matrix gelling
- Use with any TeSR™ family medium to maintain hPSCs
- Create a completely xeno-free system when used with TeSR™-E8™ or TeSR™-AOF
Support the growth and differentiation of human pluripotent stem cells by using Vitronectin XF™, a defined and xeno-free cell culture matrix.
Developed and manufactured by Nucleus Biologics, Vitronectin XF™ is an effective alternative to Corning® Matrigel®. Use Vitronectin XF™ with mTeSR™1 , mTeSR™ Plus, TeSR™-E8™, or TeSR™-AOF medium to provide a defined culture system for the maintenance of human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells under feeder-free conditions. This system allows complete control over the culture environment, resulting in more consistent cell populations and reproducible results in downstream applications.
Note: CellAdhere™ dilution buffer and non-tissue culture-treated cultureware are required for use with Vitronectin XF™ and are available for purchase separately.