PCR tubes, Corning® Thermowell® GOLD and Thermowell
Supplier: Corning
Certificates
About this item
Manufactured from ultra-pure, medical grade PP suitable for RT-PCR.
- Available with flat caps for easy piercing and marking, or domed caps for efficient heat transfer
- Ultra-thin walls of uniform thickness accelerate heat transfer for shorter cycle times and higher product yields
- 100% tested for well integrity
- Certified DNase- and RNase-free and endotoxin-safe on a lot-by-lot basis
- Autoclavable at 121 °C
- Able to withstand centrifugation to 10000×g
Tubes are designed with longer cap flanges and sealed with a full-length insertion cap to safeguard against evaporation and to prevent accidental cap opening during thermal cycling.
Delivery: Assorted pack includes the following colours: blue, green, orange, red, violet, and yellow.
Specifications
- Catalog No:
- 734-1759
- 734-1758
- 732-5583
- 732-5584
- 732-5582
- 732-5587
- 732-5588
- 732-5585
- 734-1776
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa