ContactUsLinkComponent
Venting Screw N
Venting Screw N
Catalog # PAAR104284
Supplier:  Anton Paar
New Product
Venting Screw N
Catalog # PAAR104284
Supplier:  Anton Paar
New Product
Specifications
  • Description:
    Venting screw, compatible with microwave reaction system, multiwave PRO
  • Pk:
    1
Specifications
About this item
Screw for safe and convenient closing and venting of pressure vessels NXF100 and NXQ80. Consists of a PEEK body with PTFE-TFM sealing bolt.