Silicone Hose
Supplier: NEUBERT VOLUME GLASSWAERE
New Product
Certificates
About this item
Hose made of quality silicone rubber (VMQ).
- Hardness 55 Shore A
- Food safe
- Peristaltic pumps suitable
Temperature resistant from –60 to +200 °C, short-term up to 260 °C.