Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Masks
Surgical Masks
Medical Face Mask, IIR
Medical Face Mask, IIR
SOEH1005317
:
SOEHNGEN
New Product
Medical Face Mask, IIR
SOEH1005317
:
SOEHNGEN
New Product
Type:
Individual
Colour:
White
Pk:
1
Individually wrapped
:
EN14683:2019 IIR