Male-Female Luer Lock Caps
Catalog # BDAMTP-MF-O
Supplier: BD Medical
Specifications
- Colour:Orange
- Description:Male-Female Luer lock caps, sterile
- Pk:100
Specifications
About this item
Caps for Luer lock and Luer slip. Combistoppers for female and male Luer lock.
- Individual blisters (except for 394080 and 302834 that come in a tray)
DEHP or natural rubber latex are not part of the material formulation.