EasySep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit II
Supplier: STEMCELL Technologies
Certificates
About this item
Immunomagnetic positive selection of human CD138+ cells from whole blood and bone marrow (e.g. normal or malignant plasma cells for multiple myeloma research).
- Fast and easy-to-use
- No columns required
The EasySep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit is designed to isolate CD138+ to learn how to process bone marrow samples and isolate CD138+ plasma cells to increase sensitivity in FISH testing for multiple myeloma. You can also browse our Frequently Asked Questions about isolating CD138+ plasma cells to improve FISH sensitivity.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').