ContactUsLinkComponent
EasySep™ Human Cord Blood CD34 Positive Selection Kit II
EasySep™ Human Cord Blood CD34 Positive Selection Kit II
Catalog # STMC17896RF
undefined
EasySep™ Human Cord Blood CD34 Positive Selection Kit II
Catalog # STMC17896RF
Specifications
  • Description:
    EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips, 1000 ml cord blood processed
  • Seq:
    02
  • Pk:
    1 KIT
Specifications
About this item
Complete kit for CD34+ cells from cord blood.
  • Fast and easy-to-use
  • Up to 98% purity
  • No columns required
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
Isolate highly purified CD34+ cells from fresh whole umbilical cord blood using a simple, two-step procedure. First, hematopoietic progenitor cells are pre-enriched using the RosetteSep™ Human Cord Blood CD34 Pre-Enrichment Cocktail is recommended.
If isolating CD34+ cells from other samples, including fresh or previously frozen mobilised peripheral blood or bone marrow mononuclear cells, or from previously frozen cord blood mononuclear cells, we recommend using the EasySep™ Human CD34 Positive Selection Kit. This product replaces the stand alone EasySep™ Human Cord Blood CD34 Positive Selection Kit .
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').