About this item
The BD SafetyGlide™ is designed to ensure protection from needlestick injuries after injection. Easy activation keeps healthcare workers protected and efficient.
- 'Bevel up' design eliminates safety arm interference to facilitate low angle injections
- Needle is safely protected after shielding
- Sterile, single-use design
- Natural rubber latex is not part of the material formulation
BD SafetyGlide™ safety needles with BD Activation Assist™ technology help protect from needlestick injuries through integrated safety mechanisms. BD Activation Assist™ technology ensures quick coverage of the needle by a single finger stroke immediately after injection.
The fluid, single-finger activation requires minimal training and no significant change in injection technique. The hinge arm can be pushed forward manually after use, allowing a mechanism to move forward towards the needle point. After the arm movement has been initiated manually, the hinge arm mechanism completes the full engagement. When the cover reaches the needle point it slides over the point and provides a secure encapsulation.