ContactUsLinkComponent
EasySep™ Mouse Mesenchymal Stem/Progenitor Cell Enrichment Kit
EasySep™ Mouse Mesenchymal Stem/Progenitor Cell Enrichment Kit
Catalog # STMC19771
undefined
EasySep™ Mouse Mesenchymal Stem/Progenitor Cell Enrichment Kit
Catalog # STMC19771
Specifications
  • Description:
    EasySep™ mouse mesenchymal stem/progenitor cell enrichment kit, 4×10⁸ cells
  • Pk:
    1 KIT
Specifications
About this item
Immunomagnetic isolation of untouched mouse mesenchymal stem/progenitor cells
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Isolated cells are untouched
The EasySep™ Mouse Mesenchymal Stem/Progenitor Cell Enrichment Kit is designed to isolate mesenchymal stem/progenitor cells from mouse compact bone by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies that are directed against non-mesenchymal stem/progenitor cells (CD45, TER119). labelled cells are then recognized by Tetrameric Antibody Complexes that are directed against biotin and dextran. These cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').