EasySep™ Direct HLA B Cell Isolation Kits
Supplier: STEMCELL Technologies
Certificates
About this item
Immunomagnetic isolation of untouched human B cells for HLA analysis.
The EasySep™ Direct HLA B Cell Isolation Kit can be used to isolate highly purified B cells directly from human whole blood, buffy coat, spleen, or lymph nodes by immunomagnetic negative selection.
This kit targets non-B cells for removal with antibodies recognising specific surface markers. Unwanted cells are labeled with antibodies and EasySep™ Direct RapidSpheres™, and separated using an EasySep™ magnet. Desired cells are simply collected into a new tube and can directly be used for downstream applications such as flow cytometry crossmatch.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).