
AmberChrom™ hydrogen form 50WX8 200-400 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69011-22-9
:
MDL Number:MFCD00132726
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:beige to green-beige to light brown beads
- Total exchange capacity:≥1.7 eq/l (H form)
- Water retention capacity:50 to 58 % (H form)
- Particle size:200 to 400 mesh