Order Entry
Germany
ContactUsLinkComponent
Soda lime, granular, pure, without indicator
Corrosive
Soda lime, granular, pure, without indicator
Catalog # 472160010.
CAS Number:  
Corrosive
Soda lime, granular, pure, without indicator
Catalog # 472160010.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Appearance:
    white to beige to light grey granules, pellets and/or chunks
  • Loss on drying:
    ≤20 % (10 g, 200°C, 2 h)
  • CO2 absorption capacity:
    ≥19.0 %
  • Additional info:
    Without indicator

Specification Test Results

About this item

 :
Merck Index:13,00864
Storage Temperature:Ambient
UN:1907
ADR:8,III
 :GHS05
CAS Number:8006-28-8
MDL Number:MFCD00134094