Trifluoroacetic acid 98%, pure
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
CAS Number:76-05-1
:
Formula:C₂HF₃O₂
MDL Number:MFCD00004169
:114.02 g/mol
Boiling Pt:72°C (760mmHg)
Density:1.49
Storage Temperature:Ambient
EINECS:200-989-3
UN:2699
ADR:8,I
:GHS07
:GHS05
Specification Test Results
- Appearance:colorless to light yellow clear liquid
- Infrared spectrum:Conforms
- Titration with NaOH:97.5 to 102.5 %
- Water:≤0.5 %