
Di-tert-butyloxalate 96%
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
CAS Number:691-64-5
:
Formula:C₁₀H₁₈O₄
MDL Number:MFCD00008806
:202.24 g/mol
Boiling Pt:229.3°C
EINECS:211-723-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:colorless to almost white powder or crystals
- Infrared spectrum:Conforms
- GC:≥95 %
- Melting point:69°C to 72°C