Nafamostat mesylate
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
CAS Number:82956-11-4
:
Formula:C₁₉H₁₇N₅O₂
MDL Number:MFCD00941430
:539.58 g/mol
Merck Index:13,06376
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:white to beige crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Loss on drying:≤1 %