
2-Bromo-9,10-anthraquinone
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
CAS Number:572-83-8
:
Formula:C₁₄H₇BrO₂
MDL Number:MFCD00474541
:287.11 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:light yellow to brown to dark green crystalline powder and chunks
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Melting point:205°C to 211°C