Glycerine, meets analytical specification of Ph.Eur.
New Product
:56-81-5
:
:HOCH₂CH(OH)CH₂OH
:MFCD00004722
:92.09 g/mol
:290°C (760mmHg)
:1.261
:160°C
:Ambient
:200-289-5
:Exempted from REACH
- Appearance:clear colorless to almost colorless viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4700 to 1.4750 (20°C, 589 nm)
- Specific gravity:1.249 to 1.268
- Assay:98.0 to 101.0 % (On anhydrous substance)