Sterilisation Rolls, Tyvek
Supplier: STERIS
New Product
Certificates
About this item
These sterilisation tubes from STERIS are made of Tyvek. This unique material for sterile barrier systems is known for its exceptional strength, durability and tear resistance.
- Color: White
It can be used for ethylene oxide and gamma sterilization, but also for VHP sterilization (with hydrogen peroxide vapor).
Sterilisation tubes are suitable for packaging particularly long instruments or for producing individual bags with extra length. Tyvek rolls can be sealed at 100 to 120 °C.
Applications: Department surgery, orthopedics, urology, dermatology, general medicine, dental, gynecology, orthodontics, ENT, maxillofacial, ophthalmology, veterinary, tattoo, podiatry.
Certifications: The Tyvek rolls were manufactured under quality characteristics corresponding to DIN EN ISO 13485 and comply with DIN EN ISO 11607-1 and -2.