Order Entry
Germany
ContactUsLinkComponent
 
Eriochrome Black T mixture indicator
Catalog # ACSDCH8005015.L04
Supplier:  CPACHEM
New Product
CAS Number:  
Eriochrome Black T mixture indicator
Catalog # ACSDCH8005015.L04
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    40 mL
  • Seq:
    2360

Specifications

Specification Test Results

  • Content:
    0.1g of methyl red in 7.4ml of 0.05mol/l NaOH solution and diluted with water to 200ml.
  • Performance Test:
    passed
  • Colour change:
    from red to yellow (pH 4.2 - 6.3)

Specification Test Results

About this item

 :
Melting Pt:801 °C
 :461.39 g/mol
Storage Temperature:Room Temperature
CAS Number:1787-61-7
Formula:C₂₀H₁₂N₃NaO₇S
MDL Number:MFCD00003935