Order Entry
Germany
ContactUsLinkComponent
 
BSA-Linoleate ω-6 Polyunsaturated Fatty Acid Complex
  CAYM38649-5
New Product
 :  
BSA-Linoleate ω-6 Polyunsaturated Fatty Acid Complex
  CAYM38649-5
New Product
 :  

 

  • Concentration:
    1 mM
  • Pk:
    5 mL
  • Description:
    BSA-Linoleate ω-6 polyunsaturated fatty acid complex

 

 

BSA-linoleate ω-6 polyunsaturated fatty acid complex is composed of linoleic acid and bovine serum albumin (BSA) at an approximately 6:1 molar ratio of linoleate:BSA.

  • Source: Animal/bovine
  • Formulation: 1 mM linoleate:0,17 mM BSA in 150 mM sodium chloride
  • Storage: –20 °C
  • Stability: ≥1 year

BSA-linoleate complexes can be used for efficient fatty acid delivery to cells in culture for the purpose of monitoring fatty acid oxidation or similar processes in various cellular metabolic studies. The BSA/BSA-FAs are acceptable for use with short-term cell culture applications (acute treatment to 18 hours), however, for long-term applications (25+ hours) the product can/should be filter-sterilised using a 0,2 µm filter and sterile receptacle without affecting the performance.

For best results, it is recommended that this product be used in conjunction with BSA control for BSA-fatty acid complexes (1 mM).