Order Entry
Germany
ContactUsLinkComponent
Sampling Tube
Sampling Tube
Catalog # DISI4050-0075
Supplier:  DISTEK
New Product
Sampling Tube
Catalog # DISI4050-0075
Supplier:  DISTEK
New Product

Specifications

  • Tubing ext.Ø:
    12,7 mm
  • Description:
    Sampling pipe, harvest tube, 212 mm depth for 2 and 5 L bioreactors
  • Pk:
    1
  • Hose barb size:
    Ø 12,7 mm

Specifications

About this item

Sampling tube, harvest tube for M18 port.