ContactUsLinkComponent
Molykote® High vacuum grease
Molykote® High vacuum grease
Catalog # 291-0039
Supplier:  DOW CORNING
Molykote® High vacuum grease
Catalog # 291-0039
Supplier:  DOW CORNING
Specifications
  • Description:
    High vacuum grease
  • Pack type:
    Aluminium tube
  • Pk:
    50 g
Specifications
About this item
Light grey, translucent, non melting silicone. Serviceable from –40 to +260 °C.
Delivery: This product is not available in all countries. Please check with your local Avantor sales office.