
Diphenylamine 1 g/l in sulphuric acid R Reag. Ph. Eur. 1032101
Catalog # 85826.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:169.23 g/mol
Storage Temperature:Ambient
EINECS:204-539-4
UN:1830
ADR:8,II
:GHS05
CAS Number:122-39-4
Formula:(C₆H₅)₂NH
MDL Number:MFCD00003014