Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonia solution
Ammonia solution 17% Reag. Ph. Eur. 1004701
Ammonia solution 17% Reag. Ph. Eur. 1004701
Ammonia solution 17% Reag. Ph. Eur. 1004701
87707.180
:
VWR Chemicals
:
Ammonia solution 17% Reag. Ph. Eur. 1004701
87707.180
:
VWR Chemicals
:
Pack type:
Plastic bottle
Pk:
100 mL
Seq:
2440
:
:
35.05 g/mol
:
0.92 g/cm³ (20 °C)
:
Ambient
:
215-647-6
:
2672
:
8,III
:
GHS05
:
GHS07
:
1336-21-6
:
7664-41-7
:
NH₃
:
MFCD00066650