Vanadyl sulphate hydrate
Catalog # STRE93-2334-25G
Supplier: Strem Chemicals, Inc.
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
ADR:6.1,II
:163.00 g/mol (anhy)
EINECS:248-652-7
UN:2931
:GHS06
:GHS08
:GHS07
CAS Number:123334-20-3
Formula:O₅SV·H₂O
MDL Number:MFCD00149933