Order Entry
Germany
ContactUsLinkComponent
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) 5 mg, tablets
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) 5 mg, tablets
Catalog # PIER34047
CAS Number:  
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) 5 mg, tablets
Catalog # PIER34047
CAS Number:  

Specifications

  • Pk:
    105 Tab.
  • Seq:
    0960

Specifications

About this item

 :
Melting Pt:300 °C
 :371.144 g/mol
Storage Temperature:Freezer
EINECS:224-246-5
CAS Number:4264-83-9
Formula:C₆H₄NNa₂O₆P·6H₂O
MDL Number:MFCD00066288