4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP), powder
Supplier: Uptima
Certificates
About this item
CAS Number:4264-83-9
Formula:C₆H₄NNa₂O₆P·6H₂O
MDL Number:MFCD00066288
Melting Pt:300 °C
:371.144 g/mol
Storage Temperature:Freezer
EINECS:224-246-5
Frequently Bought Together