4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP), tablets
Catalog # UPST20-106
Supplier: UPSTATE TECHNOLOGY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
Melting Pt:300 °C
:371.144 g/mol
Storage Temperature:Freezer
EINECS:224-246-5
CAS Number:4264-83-9
Formula:C₆H₄NNa₂O₆P·6H₂O
MDL Number:MFCD00066288