Order Entry
Germany
Orders LinkContactUsLinkComponent
Florisil® 100-200 mesh for column chromatography

Florisil® 100-200 mesh for column chromatography

Florisil® 100-200 mesh for column chromatography
Certificates
Florisil® 100-200 mesh for column chromatography
Florisil® 100-200 mesh for column chromatography

3 Options Available Below

Product Details & Documents
Synonyms:  Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
  • CAS Number:1343-88-0
  • Formula:MgO₃Si
  • MDL Number:MFCD00078249
  • Molecular Weight:100.38 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-681-1

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Powder
  • Particle size:
    0.1 % (80 mesh)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 137,00 €
    137,00 €
    1 * 250 g
       137,00 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 241,00 €
    241,00 €
    1 * 500 g
       241,00 €

  • Plastic drum
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 1 630,00 €
    1 630,00 €
    1 * 5 kg
       1 630,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.