Dodecyl-β-D-glucopyranoside
Catalog # MOLE21301716-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:348.48 g/mol
Storage Temperature:Ambient
EINECS:261-614-4
CAS Number:59122-55-3
Formula:C₁₈H₃₆O₆
MDL Number:MFCD00063298