Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dodecyl β-D-glucopyranoside
Dodecyl-β-D-glucopyranoside
Dodecyl-β-D-glucopyranoside
Dodecyl-β-D-glucopyranoside
Supplier:
BIOMOL RESEARCH LABORATORIES
Certificates
Expand image
Dodecyl-β-D-glucopyranoside
Supplier:
BIOMOL RESEARCH LABORATORIES
About this item
CAS Number:
59122-55-3
:
Formula:
C₁₈H₃₆O₆
MDL Number:
MFCD00063298
:
348.48 g/mol
Storage Temperature:
Ambient
EINECS:
261-614-4