4-Amino-3-hydroxy-1-naphthalenesulphonic acid, Supelco®
: Merck
:116-63-2
:
:C₁₀H₉NO₄S
:MFCD00004019
:239.25 g/mol
:13,00453
:466 °C (1013 hPa)
:∼285 °C
:M-Clarity™ quality level = MQ100
:Ambient
:204-147-3
- Solubility:(20 °C) practically insoluble
- Molar Mass:239.24 g/mol
- Bulk Density:280 kg/m3