ENERGY Easytabs for Dishwashers
Catalog # TANA715844
Supplier: WERNER MERTZ
New Product
Specifications
- Description:ENERGY easytabs, 5×85 tabs
- Pk:5
- pH value:11
Specifications
About this item
ENERGY easytabs four in one tabs for dishwashers.
- Multi-performing
- Easy dosage
- Material friendly
Detergent: Removes stains and grease and makes your dishes clean
Rinse aid: Accelerates drying time, prevents lime scale deposits, and makes dishes sparkle
Regeneration: Prevents the formation of white veil, for bright dishes
Protection: Protects your dishes, for a longer lifespan
Meanwhile enhancing the work safety, the product is phosphate free and fragrance free and prevents scaling of the heating elements.
Majorly composed of renewable resources, ENERGY easytabs takes on the responsibility for future generations.