Order Entry
Germany
ContactUsLinkComponent
 
Sucralose, powder, EMPROVE® Ph Eur, ChP, NF, JPE
Catalog # 1.00894.1000
Supplier:  Merck
New Product
CAS Number:  
Sucralose, powder, EMPROVE® Ph Eur, ChP, NF, JPE
Catalog # 1.00894.1000
Supplier:  Merck
New Product
CAS Number:  

Specifications

  • Pack type:
    Fibre carton
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay (HPLC, calc. on anhydrous substance):
    98.0 - 102.0 %
  • Assay (HPLC, calc. on anhydrous substance) (ChP):
    98.0 - 102.0 %
  • Assay (calc. on anhydrous substance) (JPE):
    98.0 - 102.0 %
  • Appearance of substance (description):
    balled powder
  • Appearance of substance (color):
    almost white
  • Identity (HPLC):
    conforms
  • Identity (TLC):
    conforms
  • Identity (IR):
    conforms
  • Identity (Spec. rotation (Ph Eur)):
    conforms
  • Spec. Rotation [α²0/D (calc. on anhydrous substance) (Ph Eur/JPE):
    +84.0 - +87.5 °
  • Spec. Rotation [α²0/D (calc. on anhydrous substance) (NF/ChP):
    +84.0 - +87.5 °
  • Appearance of solution (10 %, water):
    conforms
  • pH-Test:
    Decrease of 1 pH unit max.
  • Heavy metals (as Pb):
    ≤ 0.0010 %
  • Methanol:
    ≤ 0.1 %
  • Methanol (ChP):
    ≤ 0.1 %
  • Cyclohexane:
    ≤ 3880 ppm
  • N,N-Dimethylformamide:
    ≤ 880 ppm
  • Acetic acid:
    ≤ 5000 ppm
  • Ethyl acetate:
    ≤ 5000 ppm
  • Hydrolysis products:
    ≤ 0.1 %
  • Impurities H and I:
    ≤ 0.1 %
  • Monosaccharide-Chloride (as Fructose):
    ≤ 0.16 %
  • Related substances (Ph Eur/ChP):
    ≤ 0.5 %
  • Related substances (NF):
    ≤ 0.5 %
  • Other Disaccharide-Chlorides:
    ≤ 0.5 %
  • Triphenylphosphine oxide:
    ≤ 150 ppm
  • Residue on ignition:
    ≤ 0.7 %
  • Water (according to Karl Fischer):
    ≤ 2.0 %
  • Particle Size (d90):
    ≤ 12 µm
  • Microbiological purity (Total aerobic microbial count (TAMC)):
    ≤ 1E+03 CFU/g
  • Microbiological purity (Total yeast and mould count (TYMC)):
    ≤ 1E+02 CFU/g
  • Bile tolerant Gram negative Bacteria (absent in 1 g):
    conforms
  • Escherichia coli (absent in 1 g):
    conforms
  • Pseudomonas aeruginosa (absent in 1 g):
    conforms
  • Staphylococcus aureus (absent in 1 g):
    conforms
  • Candida albicans (absent in 1 g):
    conforms
  • Salmonella ssp (absent in 10 g):
    conforms

Specification Test Results

About this item

 :
 :397.63 g/mol
Melting Pt:114.5 °C
Density:1.62 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ500 (more info.)
Storage Temperature:Room Temperature
EINECS:259-952-2
UN:1845
CAS Number:56038-13-2
Formula:C₁₂H₁₉Cl₃O₈
MDL Number:MFCD03648615