Order Entry
Germany
ContactUsLinkComponent
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 98%
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 98%
Certificates
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 98%

Specification Test Results

  • Appearance (Color):
    Clear colorless to yellow
  • Form:
    Liquid
  • Identification (FTIR):
    Conforms
  • Refractive Index:
    1.4590-1.4630 @ 20°C
  • Assay (GC):
    ≥96.0%

Specification Test Results

Related Information